Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Lab Pack Open Head Poly Drum with 2 in. Bung Holes, 30 gal.
GREENER_CHOICE

Catalog No. 19338453 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Color:
Black
Blue
Yellow
Closure Type:
Metal Bolt Ring
Metal Lever-lock
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
19-338-453 Blue Metal Lever-lock
19-338-457 Black Metal Bolt Ring
19-338-456 Blue Metal Bolt Ring
19-338-455 Yellow Metal Bolt Ring
19-338-454 Black Metal Lever-lock
19-338-452 Yellow Metal Lever-lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-338-453 Supplier Eagle™ Supplier No. 1601MBBG2
Add to Cart
Add to Cart

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two, 2 in. holes and is ideal for safe storage, as well as handling or transportation of hazardous materials.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two, 2 in. holes
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Metal Lever-lock
Capacity (English) 30 gal.
Capacity (Metric) 113.7 L
Color Blue
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 26.38 in.
Height (Metric) 67 cm
Diameter (English) Opening 2 in. (Bung Holes)
Weight (English) 18 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.