Learn More
Eagle™ Lab Pack Open Head Poly Drum with 2 in. Bung Holes, 30 gal.
Shop All Eagle Manufacturing Products

Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- Bung lid includes two, 2 in. holes
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Specifications
Specifications
| Product Type | Drum |
| Closure Included | Yes |
| Closure Type | Metal Lever-lock |
| Capacity (English) | 30 gal. |
| Capacity (Metric) | 113.7 L |
| Color | Black |
| For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
| Material | HDPE |
| Head Style | Open |
| Height (English) | 26.38 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.