Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 55 gal.
GREENER_CHOICE

Catalog No. 19338406 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Color:
Black
Blue
Yellow
Closure Type:
Metal Bolt Ring
Metal Lever-lock
Plastic Lever-lock
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
19-338-406 Blue Metal Lever-lock
19-338-403 Blue Plastic Lever-lock
19-338-402 Yellow Plastic Lever-lock
19-338-412 Black Metal Lever-lock
19-338-405 Yellow Metal Lever-lock
19-338-414 Black Metal Bolt Ring
19-338-408 Blue Metal Bolt Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-338-406 Supplier Eagle™ Supplier No. 1655MB
Add to Cart
Add to Cart

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Metal Lever-lock
Capacity (English) 55 gal.
Capacity (Metric) 208.45 L
Color Blue
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 38.6 in.
Height (Metric) 98 cm
Diameter (English) Outer 23.75 in. (Top), 19.4 in. (Bottom)
Weight (English) 23.5 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.