Learn More
GA International Lab-TAG™ Cryogenic Labels – Cryogenic Color Rectangles for 1.5 ml Microtubes
Supplier:  GA International JTA2610LE
Encompass_Preferred
These labels are small diameter writable rectangular and round cryo labels for microtube sides and tops made with a strong cryo adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage (subject to testing of labels with intended vial cap). Specially designed to comfortably fit 0.5 ml, 0.6 ml, and 1.5 ml Eppendorf tube caps and microtubes, these waterproof, cryo labels are coated with a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. These freezer labels are supplied as either Hagaki sheets or convenient dispenser rolls, and are available in 15 individual colors, as well as packs of 15 assorted colors, for color coding and rapid identification.
Catalog No. NC0765314
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.