Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Lab Tools Kit
SDPSCI_ED

Catalog No. s65106 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S65106 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65106 Supplier United Scientific™ Supplier No. LSET6
Only null left
Add to Cart
Add to Cart

This Lab Tools Kit features an assortment of the most commonly used lab tools available as one convenient set.

The set contains one each of:

  • Test Tube Brush
  • Spatula
  • Crucible Tongs
  • Test Tube Clamp
  • Lab Scoop
  • Forceps

Specifications

Grade 5+
For Use With (Application) Tools for a variety of lab applications
Includes Test tube brush, spatula, crucible tongs, test tube clamp, lab scoop, forceps
Quantity 1 Ea.
Class Size 1 Lab Group
Product Type Laboratory Tool Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.