Learn More
Fisherbrand™ Lab Tools Set of 12
Catalog No. 13820087 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
12
Catalog No. Quantity
13-820-087 12
1 options
Catalog No. 13-820-087
Add to Cart
Add to Cart
Perfect for students, instructors and professional
Our lab tool set provides all of the commonly used accessory tools.Set includes: (1) Scoopula with (1) Wooden Handle (1) Crucible Tongs (1) Beaker Tongs (1) Spoonula (1) Semi Microspatula (1) Flexible Microspatula (1) Microspoon with Vinyl Grip (1) Test Tube Clamps(1) Scalpel Handle - Size #3(1) Scalpel Blade #10(1) Scalpel Blade #11.
  • Handy and economical assortment of commonly used lab tools
  • Covers variety of applications
  • High quality material
  • Reusable
  • Perfect for students, instructors and professional
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Quantity 12
Autoclavable Autoclavable
Description Universal lab tool kit
For Use With (Application) Various lab tasks
Size Various
Includes Various lab tools (See iamge and description)
Material Stainless Steel