Learn More
Description
Host Species: Mouse
Clone: MSA06; same as HUC1-1
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat, Pig, Horse, Cow and Dog. Others not tested.
Epitope: Not determined
Immunogen: Purified actin
Positive Control: Muscle or sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Actin, Muscle Specific Ab-6 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MSA06; same as HUC1-1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified actin |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.