Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin, Muscle Specific, Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1296R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1296R7 7 mL Prediluted Prediluted
MS1296P0 100 μL Concentrated 200μg/mL
MS1296P1ABX 100 μL Concentrated 1mg/mL
MS1296PABX 200 μL Concentrated 1mg/mL
MS1296P1 500 μL Concentrated 200μg/mL
MS1296P 1 mL Concentrated 200μg/mL
MS1296R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1296R7 Supplier Epredia Supplier No. MS1296R7

missing translation for 'validMainframeMsg'

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MSA06; same as HUC1-1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, Pig, Horse, Cow and Dog. Others not tested.

Epitope: Not determined

Immunogen: Purified actin

Positive Control: Muscle or sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Actin, Muscle Specific Ab-6
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MSA06; same as HUC1-1
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified actin
Quantity 7 mL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Equine, Human, Mouse, Pig, Rat
Form Prediluted
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.