Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Calretinin Rabbit Monoclonal Antibody

Catalog No. RM9113S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9113S 1 mL Concentrated
RM9113S0 100 μL Concentrated
RM9113S1 500 μL Concentrated
RM9113R1 1 mL Prediluted
RM9113RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9113S Supplier Epredia™ Supplier No. RM9113S

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ Calretinin, Rabbit Monoclonal Antibody.

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Rabbit

Clone: SP13

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length mouse calretinin protein

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Calretinin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP13
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant full-length mouse calretinin protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.