Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD138 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9422P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RB9422P0 100 μL
RB9422P1 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RB9422P0 Supplier Epredia™ Supplier No. RB9422P0

Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD138, Rabbit Polyclonal Antibody.

CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence is a useful marker for viable myeloma cells.

Host Species: Rabbit

Species Reactivity: Human. Others not tested

Epitope: aa 18-218

Immunogen: Recombinant protein encoding aa 18-218 human CD138

Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)

Positive Control: Tonsil

Cellular Localization: Plasma cell and Membrane

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD138
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding aa 18-218 human CD138
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.