Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD45RA Ab-3 Mouse Monoclonal Antibody, 1mL; 200μg/mL, BSA and azide

Catalog No. p-4546033 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1845R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1845R1 Supplier Epredia™ Supplier No. MS1845R1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD45RA Ab-3, Mouse Monoclonal Antibody.

CD45 or Leukocyte Common Antigen (LCA/T200) belongs to a family of membrane sialoglycoproteins with MW of 180kDa-220 kDa. Human CD45 contains three exons which encode peptide segments designated A, B and C, respectively. The differential splicing of the exons generates various isoforms. CD45RA.is a single chain glycoprotein. It is expressed on appoximately 40-50% of peripheral CD4+ T-cells, 50% of peripheral CD8+ T-cells, B-cells, and leukemic B-cell lines. T-cells expressing CD45RA and CD45RO define complementary, predominantly non-overlapping populations of resting peripheral T-cells. CD45RA are naive or virgin T-cells while, T-cells expressing CD45RO are memory T-cells.

Host Species: Mouse

Clone: 4KB5

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested

Epitope: Not determined

Immunogen: Hairy cell leukemia cells

Molecular Weight: 205-220 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD45RA Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4KB5
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Hairy cell leukemia cells
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.