Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody

Catalog No. MS1808S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1808S 1 mL Concentrated
MS1808S0 100 μL Concentrated
MS1808S1 500 μL Concentrated
MS1808R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1808S Supplier Epredia™ Supplier No. MS1808S
Only null left
Add to Cart
Add to Cart

Test for macrophages and monocytes with Epredia™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody.

CD68 glycoprotein is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: PG-M1

Isotype: IgG3/κ

Species Reactivity: Human. Others not tested

Epitope: Not determined

Immunogen: Gaucher cells

Molecular Weight: 110 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD68/Macrophage Marker Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PG-M1
Conjugate Unconjugated
Host Species Mouse
Immunogen Gaucher cells
Quantity 1 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG3 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.