Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ D4-GDI Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9042P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Purified
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9042P 1 mL Concentrated
RB9042P1 500 μL Concentrated
RB9042R1 1 mL Prediluted
RB9042R7 7 mL Prediluted
RB9042P0 100 μL Purified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB9042P Supplier Epredia™ Supplier No. RB9042P

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ D4-GDI, Rabbit Polyclonal Antibody.

D4-GDI belongs to the family of Rho-GDIs or GDP-dissociation inhibitors for the Rho/Rac family of small G proteins; consisting of RhoGDI-1, D4-GDI/RhoGDI-2 and RhoGDI-3. It inhibits the release of GDP from the Rho proteins, essential for GTP-loading and activation of GTPase activity. D4-GDI is located in the cytoplasm and in involved in the regulation of the membrane association and dissociation cycle of Rho/Rac proteins. It is known to be a substrate for caspase 3 during Fas-mediated apoptosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human D4-GDI protein

Molecular Weight: 28 kDa

Positive Control: Placenta and tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen D4-GDI
Applications Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant full-length human D4-GDI protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.