Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Gastrin Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. p-4545294 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB1459A0 100 μL Concentrated
RB1459A1 500 μL Concentrated
RB1459A 1 mL Concentrated
RB1459R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB1459A0 Supplier Epredia™ Supplier No. RB1459A0

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Gastrin Ab-1, Rabbit Polyclonal Antibody.

Gastrin, a polypeptide hormone, occurs naturally in three forms: gastrin-14, gastrin-17 and gastrin-34.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Immunogen: A synthetic peptide from human gastrin

Positive Control: Stomach

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin )

Specifications

Antigen Gastrin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen A Synthetic peptide from human gastrin
Quantity 100 μL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.