Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Human Chorionic Gonadotropin/hCG Mouse Monoclonal Antibody with BSA and azide

Catalog No. p-4541091 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Purified
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS9080P1 500 μL Concentrated
MS9080R1 1 mL Prediluted
MS9080R7 7 mL Prediluted
MS9080P0 100 μL Purified
MS9080P 1 mL Purified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS9080P1 Supplier Epredia™ Supplier No. MS9080P1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG, Mouse Monoclonal Antibody.

HCG is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).

Host Species: Mouse

Clone: SPM105

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested

Immunogen: Human hCG β

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Chorionic Gonadotropin, Human/hCG
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM105
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human hCG beta
Quantity 500 μL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.