Learn More
Epredia™ Lab Vision™ Insulin Ab-6 (INS04 + INS05) Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: INS04 + INS05
Isotype: IgG1/κ + IgG1/κ
Species Reactivity: Human, Rat and Pig. Others not tested.
Epitope: Not determined
Immunogen: Insulin protein
Molecular Weight: 6kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic Ab-6 cocktail is especially designed for sensitive detection of insulin .
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Insulin Ab-6 (INS04 plus INS05) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | INS04 + INS05 |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | Insulin protein |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.