Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Receptor (α-Subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS631B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS631B1 500 μL Biotin 200μg/mL
MS631P0 100 μL Unconjugated 200μg/mL
MS631P1 500 μL Unconjugated 200μg/mL
MS631P1ABX 100 μL Unconjugated 1mg/mL
MS631PABX 200 μL Unconjugated 1mg/mL
MS631B0 100 μL Biotin 200μg/mL
MS631B 1 mL Biotin 200μg/mL
MS631P 1 mL Unconjugated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS631B1 Supplier Epredia Supplier No. MS631B1

Epredia™ Insulin Receptor (α-Subunit) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: 83-14

Isotype: IgG2a/κ

Species Reactivity: Human. Weakly with Cow, Pig and Sheep. Does not react with rabbit, mouse and rat. Others not known.

Epitope: aa 469-592 (exon 7/8)

Immunogen: IM-9 lymphocytes followed by purified insulin receptor

Molecular Weight: 135kDa

Positive Control: IM-9 lymphocyte cells, Placenta

Cellular Localization: Membrane Ab-1 inhibits insulin binding and mimics insulin action. It is excellent for immunoprecipitation of alpha-subunit of IR.

Recommended for:

  • Immunoprecipitation
  • ELISA
  • Insulin-like Agonist
  • Inhibits (∽80%) Insulin Binding (10nM)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Insulin Receptor (alpha-Subunit) Ab-1
Applications Immunoprecipitation
Classification Monoclonal
Clone 83-14
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen IM-9 lymphocytes followed by purified insulin receptor
Quantity 500 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.