Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Negative Control for Rabbit IgG Ab-1, Epredia™

Catalog No. N100P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
N100P0 100 μL Purified with BSA and Azide
N100P1ABX 500 μL Purified w/o BSA and Azide
N100P 1 mL Purified with BSA and Azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N100P0 Supplier Epredia Supplier No. NC100P0

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.

This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.

Host Species: Rabbit

Isotype: IgG

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Negative Control for Rabbit IgG Ab-1
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified with BSA and Azide
Host Species Rabbit
Quantity 100 μL
Regulatory Status RUO
Research Discipline Negative Controls
Primary or Secondary Primary
Target Species All species
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.