Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)

Catalog No. TA999PT Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA999PT 1000 mL
TA125PT 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TA999PT Supplier Epredia™ Supplier No. TA999PT
Add to Cart
Add to Cart

Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).

  • For in vitro diagnostic use
  • Added to promote more effective washings, resulting in decreased nonspecific background staining
  • Aids reagent spreading on slides

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.

Specifications

Chemical Name or Material PBS and Tween
Concentration 20X
For Use With (Application) Anatomical Pathology
Quantity 1000 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.