Learn More
Epredia™ Lab Vision™ Placental Alkaline Phosphatase (PLAP) Rabbit Monoclonal Antibody,
Shop All Epredia ProductsDescription
Host Species: Rabbit
Clone: SP15
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant protein encoding human Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta or seminoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Placental Alk Phosphatase (PLAP) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP15 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encoding human Placental Alkaline Phosphatase (PLAP) |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.