Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Placental Alkaline Phosphatase (PLAP) Rabbit Monoclonal Antibody,

Catalog No. p-4557286 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
12 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RM9115S0 100 μL
RM9115RQ 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RM9115S0 Supplier Epredia™ Supplier No. RM9115S0

Placental Alk. Phosphatase (PLAP), Rabbit Monoclonal Ab is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal abs are more sensitive and specific than other available markers.

Reacts with a membrane-bound isoenzyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. This antibody is highly specific to PLAP and shows no cross-reaction with other isoenzymes of alkaline phosphatases. It is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA).

Host Species: Rabbit

Clone: SP15

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant protein encoding human Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta or seminoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Placental Alk Phosphatase (PLAP)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP15
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding human Placental Alkaline Phosphatase (PLAP)
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.