Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 Large T Antigen Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1832P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1832P0 100 μL 200μg/mL
MS1832P1 500 μL 200μg/mL
MS1832P 1 mL 200μg/mL
MS1832P1ABX 100 μL 1mg/mL
MS1832PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1832P0 Supplier Epredia Supplier No. MS1832P0

Epredia™ SV40 Large T Antigen Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb101; originally clone 412

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: C-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen SV40 Large T Antigen Ab-1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PAb101; same as clone 412
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen SV40 transformed mouse line B4
Quantity 100 μL
Regulatory Status RUO
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Simian virus 40 (SV40)
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.