Learn More
Epredia™ Lab Vision™ Synaptophysin (Neuroendocrine Marker) Rabbit Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Rabbit
Clone: SP11
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide of human synaptophysin
Molecular Weight: 38kDa
Positive Control: IMR5 cells, Pancreas or Pheochromocytoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | Synaptophysin (Neuroendocrine Marker) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SP11 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human synaptophysin |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Neuroscience |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.