Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Thomsen-Friedenreich Antigen Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS455R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS455R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS455R1 Supplier Epredia™ Supplier No. MS455R1

Provide accurate, reproducible results with the Epredia™ Thomsen-Friedenreich Antigen Ab-1, Mouse Monoclonal Antibody.

The Thomsen-Fridenreich antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. It is considered a pan-carcinoma marker.

Host Species: Mouse

Clone: A78-G/A7

Isotype: IgM/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Galβ1-3GalNAc

Immunogen: Neuraminidase-treated human red blood cells

Positive Control: KG1 cells, Human colorectal CA tissues

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO



Specifications

Antigen Thomsen-Friedenreich Antigen Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A78-G/A7
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Neuraminidase-treated human red blood cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Prediluted
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.