Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab Vision™ Tris Buffered Saline and Tween 20 (20X), Epredia™

Catalog No. TA999TT Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA999TT 1000 mL
TA125TT 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. TA999TT Supplier Epredia Supplier No. TA999TT
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Take advantage of a wash buffer for immunolabeling techniques with the Utilize Epredia™ Lab Vision™ Tris-Buffered Saline and Tween™ 20 (20X).

  • For in vitro diagnostic use
  • Tween 20 is added to promote more effective washings, resulting in decreased nonspecific background staining
  • Tween 20 aids reagent spreading on slides

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week

Specifications

Chemical Name or Material TBS with Tween (TBST)
Concentration 20X
For Use With (Application) Anatomical Pathology
Quantity 1000 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.