Learn More
Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X)
Shop All Epredia ProductsDescription
- 100X stock solution
- 100mM Tris-HCl buffer, pH 10.0
- Dilute 10-fold with distilled water before use
Storage Conditions: Store at 2-8°C. Stable for up to 18 months. Contains no preservatives.
Specifications
Specifications
| Content And Storage | 2° to 8°C. |
| Dilution | 10X |
| Product Type | HIER |
| Quantity | 125mL |
| For Use With (Application) | Immunohistochemistry |
| pH | 10 |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.