Learn More
Vascular Endothelial Growth Factor (VEGF) Rabbit Polyclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: N-terminus
Immunogen: A synthetic peptide derived from N-terminal of human VEGF
Molecular Weight: 19-22kDa (reduced)
Positive Control: Angiosarcoma
Cellular Localization: Cytoplasmic, cell surface and extracellular matrix This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Specifications
| Antigen | Vascular Endothelial Growth Factor (VEGF) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from N-terminal of human VEGF |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Angiogenesis |
| Primary or Secondary | Primary |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.