Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vascular Endothelial Growth Factor (VEGF) Rabbit Polyclonal Antibody, Epredia™

Catalog No. p-4559167 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
12 mL
500 μL
7 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RB9031P0 100 μL
RB9031P1 500 μL
RB9031P 1 mL
RB9031R7 7 mL
RB9031RQ 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB9031P0 Supplier Epredia Supplier No. RB9031P0A

Take advantage of one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Vascular Endothelial Growth Factor (VEGF), Rabbit Polyclonal Antibody.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. This antibody reacts with the 165, 189 and 121 amino acid splice variants of VEGF of human, to a lesser extent, mouse and rat.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human VEGF

Molecular Weight: 19-22kDa (reduced)

Positive Control: Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from N-terminal of human VEGF
Quantity 100 μL
Regulatory Status IVD
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.