Learn More
Labconco™ Add-A-Valve Installation Kit
SDP
Catalog No. 16200390 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Combination Glove Box
Catalog No. For Use With (Equipment)
16-200-390 Protector Combination Glove Box
1 options
Catalog No. 16-200-390 Supplier Labconco™ Supplier No. 5225703
Add to Cart
Add to Cart
Includes
Includes 0.688 in. dia. hole saw, arbor and instructions
For various glove boxes
Applicable products: Precise Basic Glove Boxes, Precise Controlled Atmosphere Glove Boxes, Precise HEPA-Filtered Glove Boxes, Protector Combination Glove Boxes, Protector Controlled Atmosphere Glove Boxes, Protector Filtered Glove Boxes, XPert Weigh Boxes
Specifications
For Use With (Equipment) Protector Combination Glove Box
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Glove Box Kit