Learn More
Labconco™ Add-a-Valve Kit for Precise™ Glove Boxes
SDP
Catalog No. 16108411 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box
Catalog No. For Use With (Equipment)
16-108-411 Precise Basic Glove Box
1 options
Catalog No. 16-108-411 Supplier Labconco™ Supplier No. 5225701
Add to Cart
Add to Cart
Requires
Manual ball valve, compression fitting, hose connector fitting, O-ring and nut
Includes necessary components to add a service valve to the main chamber or transfer chamber
  • Requires installation and 0.688 in. (1.75cm) dia. hole
Specifications
For Use With (Equipment) Precise Basic Glove Box
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Enclosure Kit
WARNING: Cancer - www.P65Warnings.ca.gov