Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Ammonia Trap Insert
SDP

Catalog No. 16108435 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Rotary Vane Vacuum Pump
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16-108-435 Rotary Vane Vacuum Pump
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-108-435 Supplier Labconco™ Supplier No. 7995600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Clear polypropylene canister with mounting bracket or clear polypropylene canister with stand for proper use

Protects rotary vane vacuum pump

  • For use with RapidVap™ Vacuum Evaporation System, Centrivap™ Concentrators or Glove Boxes
  • Contains impregnated carbon
  • Suitable for ammonia and low molecular weight amines

Specifications

Type Trap Insert
For Use With (Equipment) Rotary Vane Vacuum Pump
Weight (English) 4 lb.
Weight (Metric) 1.8 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.