Learn More
Labconco™ Atmospure Adapter Kit
SDP
Catalog No. 16200475 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise Controlled Atmosphere Glove Box
Protector™ Glove Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16-200-475 Protector™ Glove Box
16-200-474 Precise Controlled Atmosphere Glove Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-200-475 Supplier Labconco™ Supplier No. 5441901
Add to Cart
Add to Cart
Adapters allow 3/4 in. hose to connect to 3/8 in. fittings on glove boxes
AtmosPure 3/4 in. to 3/8 in. Adapter Kit has fittings that permit 3/4 in. ID neoprene hose included with 5441800 and 5441801 Kits (sold separately) to attach to glove box’s 3/8 in. inlet/outlets. Two fittings included with each kit. Reducer adapters for use with Labconco Protector Glove Boxes manufactured before 2011. Not required on Protector Glove Boxes manufactured after November 2010.
Specifications
For Use With (Equipment) Protector™ Glove Box
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Product Type Glove Box Kit