Learn More
Labconco™ Atmospure Cable
SDP
Catalog No. 16200477 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Atmospure 6-Pin Cable
Atmospure 9-Pin Cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16-200-477 Atmospure 9-Pin Cable
16-200-476 Atmospure 6-Pin Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-200-477 Supplier Labconco™ Supplier No. 5442100
Add to Cart
Add to Cart
Oxygen/Moisture monitor cable for use with Labconco Protector Glove Boxes
AtmosPure 9-Pin Oxygen/Moisture Monitor Cable is required for use when the AtmosPure is used in AUTO mode. Allows for automatic control of the AtmosPure blower from an accessory Oxygen or Moisture Monitor (sold separately).
Specifications
For Use With (Equipment) AtmosPure
Weight (English) 2 lb.
Weight (Metric) 0.9 Kg
Product Type Atmospure 9-Pin Cable
For Use With (Application) For use when the AtmosPure is used in AUTO mode; Allows for automatic control from an accessory monitor
WARNING: Cancer - www.P65Warnings.ca.gov