Learn More
Labconco™ Atmospure Tubing Kit
SDP
Catalog No. 16200473 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Double Width Glove Box
Single Width Glove Box
Product Type:
Atmospure Flex Tubing Kit, Double
Atmospure Flex Tubing Kit, Single
Atmospure Hard Tubing Kit, Double
Atmospure Hard Tubing Kit, Single
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
16-200-473 Double Width Glove Box Atmospure Hard Tubing Kit, Double
16-200-470 Single Width Glove Box Atmospure Flex Tubing Kit, Single
16-200-471 Double Width Glove Box Atmospure Flex Tubing Kit, Double
16-200-472 Single Width Glove Box Atmospure Hard Tubing Kit, Single
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16-200-473 Supplier Labconco™ Supplier No. 5441803
Add to Cart
Add to Cart
Tubing Kits connect AtmosPure Re-Gen Gas Purifier to glove box
AtmosPure Circulation Tubing Kit includes the plumbing to make the connection from the Labconco AtmosPure Re-Gen Gas Purifier to the glove box. Includes 3/4 in. OD stainless steel, hard-ducted tubing. For use with all double-width Protector Glove Boxes.
Specifications
For Use With (Equipment) Double Width Glove Box
Weight (English) 20 lb.
Weight (Metric) 9.1 Kg
Product Type Atmospure Hard Tubing Kit, Double