Learn More
Labconco™ Automatic Pressure Controller Gas and Vacuum Tubing Connection Kit
SDP
Catalog No. 16108369 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box
Catalog No. For Use With (Equipment)
16-108-369 Precise Basic Glove Box
1 options
Catalog No. 16-108-369 Supplier Labconco™ Supplier No. 5245200
Add to Cart
Add to Cart
For use with Precise™ Controlled Atmosphere Glove Boxes
Include the flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box.
Specifications
For Use With (Equipment) Precise Basic Glove Box
Weight (English) 20 lb.
Weight (Metric) 9.1 Kg
Product Type Enclosure Kit
WARNING: Cancer - www.P65Warnings.ca.gov