Learn More
Labconco™ California Büchner Funnel
SDP
Catalog No. 1035820 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Polyethylene
Catalog No. Material
10-358-20 Polyethylene
1 options
Catalog No. 10-358-20 Supplier Labconco™ Supplier No. 5510000
Add to Cart
Add to Cart
For use with Labconco Crude Fiber Apparatus.
Stainless steel and polyethylene, two piece Buchner Funnel is modified by the addition of a 200-mesh stainless steel screen that is firmly heat sealed into the bottom of the upper section.
Specifications
Material Polyethylene
For Use With (Application) Crude fiber apparatus
Product Type Buchner Funnel
WARNING: Cancer - www.P65Warnings.ca.gov