Learn More
Labconco™ CentriVap™ Strobe Light for CentriVap™ System
SDP
Catalog No. 1631755 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
CentriVap Centrifugal System
Catalog No. For Use With (Equipment)
16-317-55 CentriVap Centrifugal System
1 options
Catalog No. 16-317-55 Supplier Labconco™ Supplier No. 7464300
Add to Cart
Add to Cart
Includes
Mounting holster and hardware for attachment to the right side of the CentriVap concentrator
Enable user to view samples while rotor is spinning.
Enable user to view samples while rotor is spinning
  • Makes samples appear to stand still while rotor is in motion and chamber is under vacuum
Specifications
Type Strobe Light
For Use With (Equipment) CentriVap Centrifugal System
Weight (English) 2 lb.
Weight (Metric) 0.9 kg
WARNING: Cancer - www.P65Warnings.ca.gov