Learn More
Labconco™ Color-Smart Organic Filter Cells for Paramount Filtered Enclosures
SDP
Organic filter cells.
Supplier:  Labconco™ 6910400
Catalog No. 16-105-91
Only null left
Add to Cart
Description
Description
  • Type AC. Color-coded black
  • Suitable for vapors from organic solvents including acetone, ethyl acetate, toluene, and xylene
Specifications
Specifications
36, 72 in. W Paramount™ Filtered Enclosure
40.8 Kg
Organic solvents including acetone, ethyl acetate, toluene and xylene
90 lb.
Filter
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.