Learn More
Labconco™ Cupsink and Turret Kit for Basic™ 47 and 70 Hoods
GSA_VASDP
Catalog No. 1608029 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Fume Hood
Catalog No. For Use With (Equipment)
16-080-29 Fume Hood
1 options
Catalog No. 16-080-29 Supplier Labconco™ Supplier No. 4852500
Add to Cart
Add to Cart
For field installation.
Cupsink and Turret Kit, polypropylene cupsink, 3 in. x 6 in., and chrome turret mount on right side wall. Plumbing pipe not included.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Fume Hood
Weight (English) 5.0 lb.
Weight (Metric) 2.3 Kg
Includes Polypropylene Cupsink, 3 x 6 in., and Chrome Turret
For Use With (Application) Fumes
Product Line Cupsink and Turret kit
WARNING: Cancer - www.P65Warnings.ca.gov