Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Digital Chloridometer: Acid Reagent
SDP

Catalog No. 09311201 Shop All Labconco Corporation Products
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-311-201 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-311-201 Supplier Labconco™ Supplier No. 4425065

missing translation for 'validMainframeMsg'

For use with Labconco Digital Chloridometer

475mL bottle.

Specifications

Type Acid Reagent
For Use With (Equipment) Digital chloridometer
Quantity 475 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.