Learn More
Labconco™ Digital Chloridometer: Chloride Standard
SDP
Catalog No. 09311202 Shop All Labconco Corporation Products
Change view
Click to view available options
Quantity:
6/Pk.
Catalog No. Quantity
09-311-202 6/Pk.
1 options
Catalog No. 09-311-202 Supplier Labconco™ Supplier No. 4425066
For use with Labconco Digital Chloridometer
120 mL bottle.
Specifications
For Use With (Equipment) Digital chloridometer
Sample Dilution 100 mEq/L
Capacity (Metric) 120 mL
Quantity 6/Pk.