Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Drain Kit
SDP

Catalog No. 09681290 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Evidence Drying Cabinet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09-681-290 Protector Evidence Drying Cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-681-290 Supplier Labconco™ Supplier No. 3928200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labconco Drain Kit, when used with a Protector Evidence Drying Cabinet, allows you to dispose of substance easily through a direct floor connection.

  • Kit designed for use with Labconco Protector Evidence Drying Cabinet
  • Components include 10 ft. drain hose and fitting
  • Connects to floor drain


Specifications

For Use With (Equipment) Protector Evidence Drying Cabinet
Weight (English) 2 lb.
Weight (Metric) 0.9 Kg
Includes 10 ft. of Drain Hose, Fitting
Product Type Drain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.