Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Drying Chamber Valves for FreeZone™ Freeze-Dry Systems
GSA_VASDP

Catalog No. 1027312 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Freeze-Dry System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
10-273-12 Freeze-Dry System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-273-12 Supplier Labconco™ Supplier No. 7509600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Ampule adapters are available for connecting ampules to 1/2 in. valve.

Replacement valves for drying chamber or manifold ports provide leak-resisting high-vacuum seals.

  • Fabricated of neoprene body and gaskets, molded plastic knob and fittings
  • Valve 10-273-12 accommodates both 1/2 and 3/4 in. adapters
  • Serum bottles fit in valves without adapter

Order Info

Contact your Customer Service Representative for assistance in selecting the proper valve.

†Drying chambers, manifolds, and support shelves for stoppering tray dryer.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Drying Chamber Valve
Material Neoprene
For Use With (Application) Drying chambers and manifolds
For Use With (Equipment) Freeze-Dry System
Height (English) 3 in.
Height (Metric) 7.62 cm
Length (English) 2 in.
Weight (English) 1 lb.
Weight (Metric) 0.5 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.