Learn More
Labconco™ Ducting Parts: Connectors
SDP
Catalog No. 16200383 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Connector, Exhaust (Six Thimble)
Connector, Hard Duct
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16-200-383 Connector, Hard Duct
16-200-382 Connector, Exhaust (Six Thimble)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-200-383 Supplier Labconco™ Supplier No. 5440800
Add to Cart
Add to Cart
Connectors for Labconco ducts
Powder-coated steel, 6 in. diameter, for connecting the glove box to ductwork attached to a remote blower. Remote blower must be balanced with glove box's built-in blower.
Specifications
For Use With (Equipment) Hoods and Enclosures
Product Type Connector, Hard Duct