Learn More
Labconco™ Exhaust Transition Adapters for XPert™and Protector™XVS
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Manufacturer: Labconco™ 3912403
Description
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Lower Exhaust Transition Adapter made of one-piece molded polyethylene. Connects 2 in. x 10 in. rectangular to 6 in. diameter round for connection to 6 in. OD duct. For use with Labconco XPert Enclosures, Protector XVS Ventilation Stations, Protector Demonstration Hoods, and Protector Work Stations.Specifications
Lower connection, 6 in Duct | |
6 in. | |
5 lb. | |
Epoxy Coated Steel |
10 in. | |
Provide connection between the XPert™ enclosures and ductwork | |
24, 36, 48 in. XPert™ and Protector™ XVS work station | |
2.3kg |