Learn More
Labconco™ Exhaust Connection Kits for Forensic Enclosures
SDP
Catalog No. p-4366717 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Capture Portable Fuming System
Protector Evidence Drying Cabinet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09-682-732 Protector Evidence Drying Cabinet
09-681-283 Capture Portable Fuming System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-682-732 Supplier Labconco™ Supplier No. 3869600
Only null left
Add to Cart
Add to Cart
Exhaust connection kits to exhaust Labconco Forensic Enclosures.
Exhaust Connection Kit for Labconco Protector Evidence Drying Cabinet allows the cabinet to be connected to house exhaust or other exhaust system. Includes powder-coated steel connector with 5.0 in. (127 mm) diameter collar, 14 in. (36 cm) of 5.0 in. diameter (127 mm) flexible duct, and two clamps. The user-supplied exhaust system must be capable of exhausting 150 CFM (4.2 m3/min) at a vacuum of 0.5 inch of water (125 Pa).
Specifications
For Use With (Equipment) Protector Evidence Drying Cabinet
Weight (English) 10 lb.
Weight (Metric) 4.5 Kg
Includes Dry powder epoxy-coated steel connector w/5 in. diameter collar
Product Type Exhaust Connection Kit
For Use With (Application) Allows cabinet to be connected to house exhaust or other exhaust system