Learn More
Labconco™ Exterior Monitor Tubing Kits
Tubing Kits connect Oxygen or Moisture monitor to glove box
$339.00
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
16-200-480
![]() |
Labconco™
5442400 |
Exterior Oxygen Monitor Tubing Kit |
Each for $339.00
|
|
|||||
16-200-481
![]() |
Labconco™
5442500 |
Exterior Moisture Monitor Tubing Kit |
Each for $339.00
|
|
|||||
Description
Exterior Monitor Tubing Kit provides attachment tubing to allow placement of Oxygen or Moisture Monitors outside the glove box. For use with Labconco Protector and Precise Glove Boxes. For use with Moisture Monitor 5244900 amd 5244901.Specifications
| 2 lb. | |
| 5 Kg |