Learn More
Labconco™ Flask for RapidStill II
SDP
Catalog No. 09040126 Shop All Labconco Corporation Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
Catalog No. Capacity (Metric)
09-040-126 1000 mL
1 options
Catalog No. 09-040-126 Supplier Labconco™ Supplier No. 6523000
Add to Cart
Add to Cart
1000 mL round bottom, short neck, flask
1000 mL round bottom, short neck, flask
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Product Type Round Distillation Flask
Standard Taper Joint 29/42
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Rimmed
Sterility Non-sterile
Bottom Shape Round