Learn More
Labconco™ Freeze Dry Replacement Parts: Lid Gasket
SDP
Catalog No. LCN76908 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Labconco Freeze Dry Systems
Catalog No. For Use With (Equipment)
LCN-76908 Labconco Freeze Dry Systems
1 options
Catalog No. LCN-76908 Supplier Labconco™ Supplier No. 7690800
Add to Cart
Add to Cart
Gasket, Lid, Labconco; Replacement, Use on all Labconco Freeze Dry Systems
  • Replacement rubber gasket
  • Seals chamber lid of FreeZone Freeze Dryer during lyophilization process
  • Creates tight barrier preventing leakage
Specifications
Type Lid Gasket
No. of Shelves 0
Material Rubber
For Use With (Equipment) Labconco Freeze Dry Systems