Learn More
Labconco™ G4 Prefilter Starter Kits
For use with Purifier™ Logic™-plus biosafety cabinets
Supplier: Labconco™ 3406000
Description
- Includes pleated disposable filter with Merv 8 rating, hardware and bracket
Installation is required.
Specifications
| G4 Prefilter | |
| 3 lb. | |
| Pleated disposable filter with Merv 8 rating, hardware and bracket |
| Purifier Logic Biosafety Cabinet (3 ft.) | |
| 1.4 kg | |
| Prefilter Kit |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov